Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse VEGF-121, Recombinant Protein, Invitrogen™

Recombinant Protein

Manufacturer:  Invitrogen RP8674

Catalog No. PIRP8674

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Vascular Endothelial Growth Factor 121 (mouse) Recombinant Protein, RP-8674, from Invitrogen.

Vascular Endothelial Growth Factor-A (VEGF-A) was originally isolated from tumor cells and is produced by a wide variety of cell types. In addition to stimulating vascular growth and vascular permeability, VEGF-A may play a role in stimulating vasodilation via nitric oxide-dependent pathways. Mouse VEGF-A has several variants, one being VEGF-120. Rat and bovine VEGF are one amino acid shorter than the human factor, and the bovine and human sequences show a homology of 95%. Recombinant mouse EGF is a non-glycosylated protein, containing 53 amino acids, including 3 intra-molecular disulfide-bonds, with a molecular weight of 6 kDa. Purity is typically greater than 97% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the dose-dependent proliferation of mouse BALB/c 3T3 cells and is typically less than 0.1 ng/mL. The specific activity of this protein is 1x10^7 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: NSYPGCPSSY DGYCLNGGVC MHIESLDSYT CNCVIGYSGD RCQTRDLRWW ELR.
Specifications

Specifications

Q00731
Control
Control
water with no preservative
28.4
purified
E. coli
RUO
Mouse VEGF-121
Mouse
Ms
Recombinant
Recombinant
0.1mg/mL
Lyophilized
22339
Mouse VEGF-121
10μg
-20°C, Avoid Freeze/Thaw Cycles
M-VEGF-121
Vegfa
Unlabeled
E. coli
Safety and Handling

Safety and Handling

ShelfLife : 12 Months

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

in vitro research only.