Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Prism RPN Pioneer HPLC Columns
Supplier: Thermo Scientific™ 32005102155
Description
Specifications
2.1 mm | |
5 μm | |
Alkyl Amide | |
No |
100 mm | |
L60 | |
Polar Embedded Silica |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction