Learn More
Thermo Scientific Remel Acetamide Agar
Thermo Scientific Remel Acetamide Agar is used for differentiation of nonfermentative gram-negative bacteria, especially Pseudomonas aeruginosa.
Manufacturer: Thermo Scientific R060010
This item is not returnable.
View return policy
Description
Thermo Scientific Remel Acetamide Agar is used for differentiation of nonfermentative gram-negative bacteria, especially Pseudomonas aeruginosa.
Specifications
Acetamide Agar | |
Agar | |
15mm x 103mm Tube |
Agar | |
20 x Agar Slant | |
20/Pk. |