Learn More
Thermo Scientific™ Remel™ BactiSwab™ Liquid Collection and Transport Systems
Provide excellent recovery in routine bacteriology and is compatible with many rapid screening methods.
$1111.00
Specifications
For Use With (Application) Provides excellent recovery in routine bacteriology and is compatible with many rapid screening methods
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Shaft Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Shaft Price Quantity & Availability  
R12702
View Documents
Thermo Scientific™
R12702
1000 Case One Plastic shaft, one Stainless Steel shaft
Case of 1000 for $1,111.00
Add to cart
Description
Description
Rayon-tipped, plastic-shaft swabs are mounted in the cap of a translucent plastic tube with spiral chamber. No transport media.
Available in several formats:
  • Single swabs with single plastic shaft, white cap
  • Double swabs with double plastic shaft, red cap
  • Liquid Stuarts, Wire Shaft, with mini-tip swab suitable for ear, nose, throat, eye, male urethral, and pediatric specimen collection; straight aluminum-shaft; green cap
Specifications
Specifications
Provides excellent recovery in routine bacteriology and is compatible with many rapid screening methods