Learn More
Thermo Scientific™ Remel™ Fluid Thioglycollate Medium w/indicator, dextrose
Fluid Thioglycollate Medium; General purpose liquid medium for cultivation of aerobes, anaerobes, microaerophilic organisms
Supplier:  Thermo Scientific™ R119136
Catalog No. R119136
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • For sterility testing according to USP (United States Pharmacopeia)
Compliance
Industrial Reference: AOAC, BAM, COMPF, USP.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 mL
100/Pk
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
REF numbers beginning with R11 are not intended for IVD use.