Learn More
Thermo Scientific™ Remel™ Pseudomonas F Agar (FLO Agar) Available on GSA/VA Contract for Federal Government customers only.
Use for the differentiation of Pseudomonas aeruginosa from other Pseudomonas species based on fluorescein production.
Manufacturer:  Thermo Scientific™ R062632
 View more versions of this product
Catalog No. R062632
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Thermo Scientific™ Remel Pseudomonas F Agar (FLO Agar) is used for the differentiation of Pseudomonas aeruginosa from other Pseudomonas species based on fluorescein production.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Pseudomonas F Agar (FLO Agar)
Agar
15mm x 103mm Tube
20/Pk.
Agar
20 x Agar Slant
Industrial Reference: BAM, USP.