Learn More
Thermo Scientific Remel Tetrathionate Broth
Thermo Scientific Remel Tetrathionate Broth is used for selective enrichment and isolation of Salmonella species.
Manufacturer: Thermo Scientific R112634
This item is not returnable.
View return policy
Description
Thermo Scientific Remel Tetrathionate Broth is used for selective enrichment and isolation of Salmonella species.
Specifications
Tetrathionate Broth | |
Broth | |
BB - 1000mL | |
4/Pk. |
Broth | |
4 x 1000mL | |
Industrial Reference: AOAC, BAM, COMPF, SMWW, USDA, USP. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Requires addition of Iodine Solution, REF R21228, REF R114350, or REF R114351. REF R112634 and REF R112630 not intended for IVD use.