Thermo Scientific™ Revco Freezer Low-Voltage Safeguards
Manufacturer: Thermo Scientific™ 55753D
 View more versions of this product
Catalog No. 13-990-46B
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Part Not Found  Please contact customer service at 1-800-766-7000 with error code:999. 
    Specifications
    For Use With Thermo Scientific™ Revco™ Elite™ and Value™ freezer
    Hertz 60Hz
    Voltage 208/230V
    Electrical Requirements 208/230V 60Hz
    Product Type Low Voltage Safeguard
    For Thermo Scientific Revco Elite™ and Value™ Freezers
    • Helps protect electrical equipment from damage by automatically boosting low voltage
    • Eliminates start-up problems
    • Provided with terminal board for direct wiring
  • Description & Specifications
Related Products
Product Content Feedback