Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Revco™ High-Performance Laboratory Refrigerators with Glass Doors
Cabinet sizes to fit your space and storage needs
Revco high-performance lab refrigerators with glass doors are available in a wide range of cabinet sizes to fit virtually any application need. Each refrigerator comes with an advanced microprocessor temperature control, high-quality cabinet construction and advanced alarm systems. Postivie, forced-air circulation maintains temperature uniformity, protecting your valuable samples and inventory.
$5394.00 - $13367.40
Specifications
Material Scratch-resistant Painted
Surface Coating High-impact powder paint coating
Metric Temperature Range 1° to 8°C
Defrost Type Automatic
Insulation Type CFC-free
View More Specs
  • Products 26
  • Description & Specifications
Products
Catalog Number Mfr. No. Description Price Quantity Availability  
RGL404A
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
RGL404A
4.9 cu. ft.; Glass door, 115V 60Hz
Each for $5,394.00
Check Availability
Add to Cart
RGL404D
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
RGL404D
4.9 cu. ft.; Glass door, NEMA 6-15P plug type; 208/230V 60Hz
Each for $5,394.00
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL404V
Thermo Scientific
RGL404V
4.9 cu. ft.; Glass door, European plug type; 230V 50Hz
Each for $5,394.00
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL404W
Thermo Scientific
RGL404W
4.9 cu. ft.; Glass door, U.K. plug type; 230V 50Hz
Each for $5,394.00
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL1204A
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
RGL1204A
11.5 cu. ft.; Glass door, 115V 60Hz
Each for $5,858.00
Check Availability
Add to Cart
RGL1204D
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
RGL1204D
11.5 cu. ft.; Glass door, NEMA 6-15P plug type; 208/230V 60Hz
Each for $5,858.00
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL1204V
Thermo Scientific
RGL1204V
11.5 cu. ft.; Glass door, European plug type; 230V 50Hz
Each for $5,858.00
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL2304W
Thermo Scientific
RGL2304W
23CF REF GLASS 230V23CF REF G
Each for $6,496.00
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL1204W
Thermo Scientific
RGL1204W
11.5 cu. ft.; Glass door, U.K. plug type; 230V 50Hz
Each for $5,858.00
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL2304A
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
RGL2304A
23.3 cu. ft.; Glass door, 115V 60Hz
Each for $6,496.00
Check Availability
Add to Cart
RGL2304D
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
RGL2304D
23.3 cu. ft.; Glass door, NEMA 6-15P plug type; 208/230V, 60Hz
Each for $6,496.00
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL2304V
Thermo Scientific
RGL2304V
23.3 cu. ft.; Glass door, European plug type; 230V 50Hz
Each for $6,496.00
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL3004A
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
RGL3004A
29.9 cu. ft.; Glass door, 115V 60Hz
Each for $7,499.00
Check Availability
Add to Cart
RGL3004D
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
RGL3004D
29.9 cu. ft.; Glass door, NEMA 6-15P plug type; 208/230V 60Hz
Each for $7,499.00
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL3004V
Thermo Scientific
RGL3004V
29.9 cu. ft; Glass door, European plug type; 230V 50Hz
Each for $7,499.00
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL3004W
Thermo Scientific
RGL3004W
29.9 cu. ft; Glass door, U.K. plug type; 230V 50Hz
Each for $7,499.00
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
13-991-139
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
REL4504A
45.8 cu. ft.; 2 sliding glass doors, 115V 60Hz
Each for $8,580.00
Check Availability
Add to Cart
13-991-140
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
REL4504D
45.8 cu. ft.; 2 sliding glass doors, 230V 50Hz
Each for $8,580.00
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL5004A
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
RGL5004A
51.1 cu. ft.; 2 glass doors, 115V 60Hz
Each for $9,747.00
Check Availability
Add to Cart
RGL5004D
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
RGL5004D
51.1 cu. ft.; 2 glass doors, NEMA 6-15P plug type; 208/230V 60Hz
Each for $9,747.00
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL5004V
Thermo Scientific
RGL5004V
51.1 cu. ft.; 2 glass doors, European plug type; 230V 50Hz
Each for $9,747.00
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL5004W
Thermo Scientific
RGL5004W
51.1 cu.ft; Glass door, U.K. plug type; 230V 50Hz
Each for $9,747.00
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL7504A
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
RGL7504A
78.8 cu. ft.; Glass door, 115V 60Hz
Each for $13,367.40
Check Availability
Add to Cart
RGL7504D
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
RGL7504D
78.8 cu. ft.; Glass door, NEMA 6-15P plug type; 208/230V 60Hz
Each for $12,405.00
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL7504V
Thermo Scientific
RGL7504V
78.8 cu. ft.; Glass door, European plug type; 230V 50Hz
Each for $12,405.00
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL7504W
Thermo Scientific
RGL7504W
78.8 cu. ft.; Glass door, U.K. plug type; 230V 50Hz
Each for $12,405.00
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Descriptions & Specifications
Related Products