Learn More
Epredia™ Richard-Allan HRP/DAB Immunohistochemistry Detection Kits
Supplier: Epredia™ R5HD
This item is not returnable.
View return policy
Description
- Hydrogen Peroxide
- Protein Block
- Biotinylated Anti-mouse/rabbit lgG
- Streptavidin HRP
- DAB Buffer
- DAB Chromogen
Specifications
5 mL |