Learn More
Thermo Scientific™ Richard-Allan HRP/DAB Immunohistochemistry Detection Kits
Products 2
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
22-110-142
View Documents
Thermo Scientific™
R15HD
N/A HRP/DAB This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
22-110-141
View Documents
Thermo Scientific™
R5HD
N/A HRP/DAB This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Hydrogen Peroxide
  • Protein Block
  • Biotinylated Anti-mouse/rabbit lgG
  • Streptavidin HRP
  • DAB Buffer
  • DAB Chromogen