Thermo Scientific™ Rotors for Savant™ DNA SpeedVac™ Concentrators Available on GSA/VA Contract for Federal Government customers only.
Accessory Rotors
Manufacturer: Thermo Scientific™ RH625
 View more versions of this product
Catalog No. 20-548-117
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Holds 6 x 30mL Tube
    Capacity (Metric) 30mL
    For Use With (Equipment) Savant™ DNA SpeedVac™ concentrator rotor
    Product Type Glass And Plastic Tube
    Total Count 6.0
    Easy to use circular rotors with different tube hole sizes to accommodate different sample sizes. These rotors require no additional accessories and fit directly into your concentrator.
  • Description & Specifications
Related Products
Product Content Feedback