Thermo Scientific™ Samco™ Bio-Tite™ Urine Specimen Containers

Easy, stackable storage

Manufacturer: Thermo Scientific™ 02 0147

 View more versions of this product

Catalog No. 13-711-72

  Check Availability
Add to Cart

    Specifications

    Product Type Specimen Container
    Color Clear
    Sterility No
    Closure Color Yellow
    Capacity (English) 3 oz.
    Closure Type Dual-threaded cap
    For Use With (Application) For stackable storage
    Capacity (Metric) 90mL
    Capacity 3 oz., 90mL
    Material Virgin Polypropylene

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit