Learn More
Thermo Scientific™ Savant SPD1010 and SPD2010 SpeedVac Kits
For HPLC fractions with water/0.1% trifluoroacetic acid and reversed-phase SPE eluants applications
Manufacturer:  Thermo Scientific™ SPD1010A115
Catalog No. 11-716-812
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
All of our SpeedVac kits are complete systems, containing everyting you need under one model number. Our Savant SPD1010 and SPD2010 models fully incorporate four individual components: concentrator bowl, refrigerated trap, vacuum gauge and a diaphragm pump.
  • Easily programmable
  • Integrated design for plug-in simplicity and control
  • Digital display for viewing and setting parameters (time, temperature vacuum)
  • Vacuum control for vacuum ramping and ultimate level settinig to optimize drying times and eliminate bumping of different solvent mixtures
  • All-in-one design makes setup and installation easy
  • Dual timers for independent control of heater and run time
  • Oil-free vacuum source eliminates the need to add/change oil
  • Standard PTFE-coated chamber
  • Radiant cover for chemical resistance and added heat source (enhanced drying times
  • Standard glass safety lid
Solvent and Solvent Combinations - SPD1010
  • Acetonitrile
  • Ethanol
  • Methanol
  • Methylene Chloride
  • Water
  • Acetonitrile and Water
  • Methanol and Water
Solvent and Solvent Combinations - SPD2010
  • Acetonitrile
  • Ethanol
  • Methanol
  • Water
  • Acetonitrile and Water
  • Ethanol and Water
  • Methanol and Water
  • Choose the SPD1010 for capacities up to 100mL
  • Choose the SPD2010 for capacities up to 500mL
  • HPLC fractions with water/0.1% trifluoracetic acid
  • Reversed-phase SPE eluants
Specifications
Specifications
4L
60Hz
40 x 1.5mL tubes
115V
115V 60Hz