Thermo Scientific™ Screw-top Sterile Coliform Water Sample Bottle; Polystyrene, polyethylene, or polypropylene bottles with screw closure
Meets EPA Requirement for Bact-T sampling
$375.58 - $883.04
Specifications
Color Clear
Closure Size 38-400mm
Certifications Certified Sterile; manufactured under an ISO certified quality management system.
View More Specs
Includes: Bottles assembled with caps
  • Products 3
  • Description & Specifications
Products
Catalog Number Mfr. No. Material English Outside Diameter English Height Quantity Price Quantity & Availability    
12-101-509
Thermo Scientific
P156120ST
Polystyrene 1.9 in. 3.7 in. 200 Case of 200 for $437.30
Check Availability
Add to Cart
12-101-508
Thermo Scientific
P156120PP
Polypropylene 1.8 in. 3.7 in. 200 Case of 200 for $375.58
Check Availability
Add to Cart
12-100-274
Thermo Scientific
P156-150ST
Polystyrene 1.9 in. 4.1 in. 342 Case of 345 for $883.04
Check Availability
Add to Cart
Descriptions & Specifications
Related Products