Thermo Scientific™ Section Transfer System
Consists of a special disposable blade carrier with laminar flow of water between cutting edge and heated water bath mounted directly on the microtome
Manufacturer: Thermo Scientific™ 771190
 View all options for this product
Catalog No. 77-119-0
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Type Section Transfer System
    Hertz 50/60Hz
    Voltage 220/230V
    Electrical Requirements 220/230V 50/60Hz
    Disposable Yes
    • Section stretching occurs immediately as a result of water surface tension, simultaneous with cutting
    • Water bridge transports paraffin ribbons captured and automatically transfers them into the water bath
    • A separate removable waste tray collects unwanted sections and trim debris
  • Description & Specifications
Related Products
Product Content Feedback