Learn More
Thermo Scientific™ Septa Available on GSA/VA Contract for Federal Government customers only.
A wide range to cover most applications
Manufacturer:  Thermo Scientific™ 31303210
 View more versions of this product
Catalog No. 03-050-302
  Check Availability
Add to cart
Description
Description
  • Variety of materials and temperature limits for all applications
  • For use in Thermo Scientific TRACE™ and FOCUS™ GCs and instruments from other manufacturers
  • Contaminant-free in individual blister packs
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.