Thermo Scientific™ Shandon™ Downdraft Anatomy Dissecting Table
Two hinged covers totally enclose the work area and the cadaver
Designed to provide a ventilated workstation for animal necropsies, surgeries, perfusions, and dissections. Table functions to pull fumes down and away from the user's face when connected to a negative air source, providing a healthier, safer work environment.
$18993.20
Includes: Removable 20 x 82 in. (50.8 x 208.3cm) body/specimen tray
  • Products 1
  • Description & Specifications
Products
Catalog Number Mfr. No. Description Price Quantity Availability  
23-245-557
Thermo Scientific
B1020021
Downdraft Anatomy Dissection Table
Each for $18,993.20
Check Availability
Add to Cart
Descriptions & Specifications
Related Products