Learn More
Epredia™ Shandon™ Embedding Cassettes II in Tube Packs
Compatible with Epredia™ Shandon™ MicroWriter™ cassette labelers
Specifications
Quantity 25 Cassettes x 30 Tubes
View More Specs
Products 6
Catalog Number Mfr. No. Color Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Color Quantity Price Quantity & Availability  
B1000729AQ7
View Documents
Epredia™
B1000729AQ7
Aqua 25 Cassettes x 30 Tubes Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
B1000729PE7
View Documents
Epredia™
B1000729PE7
Peach 25 Cassettes x 30 Tubes Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
B1000729LI7
View Documents
Epredia™
B1000729LI7
Lilac 25 Cassettes x 30 Tubes Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
B1000729YW7
View Documents
Epredia™
B1000729YW7
Yellow 25 Cassettes x 30 Tubes Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
B1000729BL7
View Documents
Epredia™
B1000729BL7
Blue 25 Cassettes x 30 Tubes Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
B1000729GR7
View Documents
Epredia™
B1000729GR7
Green 25 Cassettes x 30 Tubes Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description
Description
  • Designed to hold tissue specimens during processing and embedding
  • Acetal polymer cassettes with writing face at 45° angle
  • Pore style: Slotted
  • Locking lid attached by two hinges
Specifications
Specifications
25 Cassettes x 30 Tubes