Thermo Scientific™ Shandon™ Glass Coverplates
For use on Shandon Sequenza™ and Cadenza™ Immunostainers
Manufacturer: Thermo Scientific™ 72110017
 View more versions of this product
Catalog No. 72-110-017
  Check Availability
Add to Cart
    Specifications
    Product Type Coverplate
    Quantity 25/pk.
  • Description & Specifications
Related Products
Product Content Feedback