Learn More
Thermo Scientific™ Shandon™ Mucolexx™ Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Thermo Scientific™ 9990375
 View more versions of this product
Catalog No. 9990375
  Check Availability
Add to cart
Specifications
Description
Cell Preparation and Collection Fluids; Thermo Scientific Shandon; Mucolexx Mucoliquefying Fluid; Liquifies (with Agitation) Mucoid Cytology Specimens and Provides Short Term Preservation of Specimens
Description & Specifications
Specifications
Shandon Mucolexx
Light Red
Provides short term preservation of specimens
50 x 120mL, filled to 60mL
Liquid
Cups