Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Shandon™ Pathcentre™ Tissue Processor Accessories
For use with the Epredia™ Shandon™ Tissue Processor
Supplier: Epredia™ 75210161
Specifications
Wax Disposal Bag | |
Tissue Processor | |
10 Pk. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title