Thermo Scientific™ Shandon™ Tissue-Loc™ Tissue Cassettes IV
Compatible with Thermo Scientific Shandon MicroWriter™ and PrintMate™ cassette labelers
Manufacturer: Thermo Scientific™ 41722
 View more versions of this product
Catalog No. 41-722
  Check Availability
Add to Cart
    Specifications
    Pore Style Small Slots
    Closure Type Hinged
    Color Yellow
  • Description & Specifications
Related Products
Product Content Feedback