Thermo Scientific™ Shandon™ Tissue-Loc™ Tissue Cassettes IV
Compatible with Thermo Scientific Shandon MicroWriter™ and PrintMate™ cassette labelers
Manufacturer: Thermo Scientific™ 41723
 View more versions of this product
Catalog No. 41-723
  Check Availability
Add to Cart
    Specifications
    Pore Style Small Slots
    Closure Type Hinged
    Color Green
    • Disposable
    • Holds tissue specimens during embedding, processing
    • Pore style: Small slots
    • Supplied in box of 250 cassettes with attached lids
  • Description & Specifications
Related Products
Product Content Feedback