Learn More
Thermo Scientific™ Shandon™ TPX Single Sample Chamber, Caps, and Filter Cards
For use with disposable white filter cards for sample volumes up to 0.5mL and for use with disposable brown filter cards for sample volumes of 0.4mL or less
Manufacturer:  Thermo Scientific™ A78710018
 View more versions of this product
Catalog No. A78710018