Learn More
Thermo Scientific™ Shandon™ Uni-Capsettes
Hold tissue specimens during processing and embedding
Manufacturer:  Thermo Scientific™ 9996420
 View more versions of this product
Catalog No. 99-964-20
Add to cart
Description
Description
  • Pore style: small slots
  • Supplied in bags of 500 cassettes with lids packaged separately
  • Available in 10 colors
Specifications
Specifications
Slots
Separate Lid
White
Processing Cassette