Learn More
Thermo Scientific™ Shandon™ Uni-Capsettes
Hold tissue specimens during processing and embedding
Manufacturer:  Thermo Scientific™ 9996490
 View more versions of this product
Catalog No. 99-964-90
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Pore style: small slots
  • Supplied in bags of 500 cassettes with lids packaged separately
  • Available in 10 colors
Specifications
Specifications
Slots
Separate Lid
Tan
Processing Cassette