Thermo Scientific™ Shandon™ Uni-Capsettes
Hold tissue specimens during processing and embedding
Manufacturer: Thermo Scientific™ 9996494
 View all options for this product
Catalog No. 99-964-94
  Check Availability
Add to Cart
    Pore Style Slots
    Closure Type Separate Lid
    Color Lilac
    Description Processing Cassette
    • Pore style: small slots
    • Supplied in bags of 500 cassettes with lids packaged separately
    • Available in 10 colors
  • Description & Specifications
Related Products
Product Content Feedback