Learn More
Thermo Scientific™ SLIPFREE™ Connectors, Double
Universal self-adjusting connections
Manufacturer:  Thermo Scientific™ 30110
 View more versions of this product
Catalog No. 03-260-180
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Void-free and leak-free by pushing tubing and ferrule into the end-fitting
  • Compatible with all column end-fittings
  • Stainless-steel threads
  • Fingertight connections to 10,000psi
  • Choose the double configuration for frequent connections and disconnections between columns and injectors
Specifications
Specifications
0.005 in.
10cm