Learn More
Thermo Scientific™ SmartScan Solo Single Tube Reader
A compact, high-speed single tube reader unit designed for benchtop 2D barcode reading
Supplier: Thermo Scientific™ AB1750
Description
The unit can be used with all of Thermo Scientific ABgene™s 2D barcoded tubes, including the 2mL CryoCode™ tube.
- Less than 1 sec. read time
- Keyboard wedge data export option, allows result to be directly imported into any open Microsoft Windows™ application on your desktop
- Other data export option supplied as standard - ODBC, Excel™ spreadsheet, Virtual COM port
- Data export and unit read rate can be configured with a wide range of options making it easy to integrate into your workflow
- 1D and 2D barcode decode
Specifications
Single Tube Reader | |
Benchtop 2d barcode reading | |
1/Cs. |