Thermo Scientific™ Sorvall™ Adapters for Stratos™ Four-Place Swinging Bucket Rotor Available on GSA/VA Contract for Federal Government customers only.

For use with Thermo Scientific Sorvall Stratos Centrifuges in Four-Place Swinging Bucket Rotor 75-003-047 with Round Bucket 75-008-172

Manufacturer: Thermo Scientific™ 75008185

 View more versions of this product

Catalog No. 75-008-185

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) 4 x 180 Swing bucket rotor
    Color White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit