Thermo Scientific™ Sorvall™ Four-Place Swinging Bucket Rotor Available on GSA/VA Contract for Federal Government customers only.
Stainless-steel rotor body only
Manufacturer: Thermo Scientific™ 75006445
 View more versions of this product
Catalog No. 75-006-445
  Check Availability
Add to Cart
    Specifications
    Type Swinging bucket
    Autoclavable Yes
    Capacity (Metric) 4 x 250/750mL
    Includes Tool
  • Description & Specifications
Related Products
Product Content Feedback