Learn More
Thermo Scientific™ Sorvall™ Legend™ Micro 21 Microcentrifuge and Rotor Packages Available on GSA/VA Contract for Federal Government customers only.
Powerful units deliver exceptional capacity, performance and safety in a compact, user-friendly design
Manufacturer:  Thermo Scientific™ 75772488
 View more versions of this product
Catalog No. 75-77-2488
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
24 x 1.5/2mL
21,000/21,999 x G
120V
22.5cm
25.2cm
1 to 99 min., ±1 min. increments
24.3cm
CSA certified, CE IVD compliant, Certified Biosafety; IEC 61010-1, IEC 61010-2, IEC 61010-020
13.75 x 9.63 x 8.88 in. (25.2 x 24.3 x 22.5cm)
10.5kg
14,000 to 14,999rpm
No
14,800rpm
60Hz
8.88 in.
13.75 in.
<56dBA
9.63 in.
-9° to +40°C, ±1°
120V 60Hz
23 lb.
21,000 to 21,999 x G
Yes
acceleration up to 21,000 x g in 12 seconds
Promotions
Promotions
  • Axygen Scientific , Thermo Scientific
    Maximize your separation capabilities and your budget with three ways to save on Thermo Scientific™ centrifuges. Explore the offers below to find the right solution for your lab.
    01-31-2018