Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Tube Adapters for Round Rotor Buckets Available on GSA/VA Contract for Federal Government customers only.

For use with four-place rotors 75006445 and 75006434 when equipped with round bucket 75006441

Manufacturer: Thermo Scientific™ 75008134

 View more versions of this product

Catalog No. 75-008-134

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) LH-4000W and LH-4000 rotor with round bucket
    Color Light grey

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit