Learn More
Thermo Scientific™ Sorvall™ Legend™ T1/RT1 Fixed-Angle Rotors Available on GSA/VA Contract for Federal Government customers only.
For use with Legend T1/RT1 centrifuges
Manufacturer:  Thermo Scientific™ 11175741
 View more versions of this product
Catalog No. 05-376-187
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Sealed rotor
8.4cm
14,600rpm
CL30 centrifuge
6500rpm
3.3 in.
24 x 2mL
45°
3967