Learn More
Thermo Scientific™ Sorvall™ Legend™ T1/RT1 Microplate Rotor Available on GSA/VA Contract for Federal Government customers only.
For use with Legend T1/RT1 centrifuges
Manufacturer:  Thermo Scientific™ 11175631
 View more versions of this product
Catalog No. 05-376-164
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Sealed plate carrier
4,100rpm
2,160xG
2 x3 microplates or 2 x 1 filter blocks
T20 microplate rotor