Learn More
Thermo Scientific™ Sorvall™ Legend™ T1/RT1 Swinging Bucket Rotors Available on GSA/VA Contract for Federal Government customers only.
For use with Legend T1/RT1 centrifuges
Manufacturer:  Thermo Scientific™ 11175611
 View more versions of this product
Catalog No. 05-376-105
  Check Availability
Add to cart
Requires : Buckets (order separately)
Specifications Description & Specifications
Specifications
Bucket
200mL
S41 swing out rotor
Rotor body only
4,100rpm
3,026xG