Thermo Scientific™ Sorvall™ Legend™ T3/RT3 Swinging Bucket Rotor Available on GSA/VA Contract for Federal Government customers only.

Versatile high-throughput rotor for use with Legend™ T3 and RT3 centrifuges

Manufacturer: Thermo Scientific™ 11174223

 View more versions of this product

Catalog No. 05-376-292

  Check Availability
Add to Cart

Required Accessories : Buckets or carriers (order separately)

    Specifications

    Type Sealed plate carrier
    Capacity (Metric) 4 x 750mL
    Speed 4,000rpm
    Includes Rotor body only

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit