Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trajan™ Thermo Scientific SPME Liner
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Supplier: Trajan™ 092148
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Thermo Scientific instruments
Specifications
GC Inlet Liner, SPME Liner | |
5/Pk. | |
8 mm |
105 mm | |
0.8 mm | |
Thermo Scientific Trace and Focus GCs |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction