Thermo Scientific™ SUN-SRi™ Caps for 8-425 Screw Thread Vials Available on GSA/VA Contract for Federal Government customers only.
Skirted Shimadzu™, in multiple colors
Manufacturer: Thermo Scientific™ 500 066
 View more versions of this product
Catalog No. 14-823-307
  Check Availability
Add to Cart
    Specifications
    Pre-slit No
    Material Polypropylene
    Type Screw Cap
    Material Closure Polypropylene
    Material (Liner) PTFE/Silicone
    Thread Size 8-425
    Color Black
  • Description & Specifications
Related Products
Product Content Feedback