Thermo Scientific™ SUN-SRi™ Centrifugal Filter Devices Available on GSA/VA Contract for Federal Government customers only.
Combination filter/receiver tube unit for sample processing directly in centifuges
Manufacturer: Thermo Scientific™ 64520NP
 View more versions of this product
Catalog No. 10-800-199
  Check Availability
Add to Cart
    Specifications
    Housing Material Polypropylene
    Max. Force 5,000xG
    Volume (Metric) Up to 2mL
    For Use With Benchtop and floor model centrifuges
    Membrane Material PTFE
    Pore Size 0.45μm
    Includes Filter housing; membrane; receiver tube
  • Description & Specifications
Related Products
Product Content Feedback