Thermo Scientific™ SUN-SRi™ Centrifugal Filter Devices Available on GSA/VA Contract for Federal Government customers only.
Combination filter/receiver tube unit for sample processing directly in centifuges
Manufacturer: Thermo Scientific™ 62215CA
 View more versions of this product
Catalog No. 10-800-184
  Check Availability
Add to Cart
    Specifications
    Housing Material Polypropylene
    Includes Filter housing; membrane; receiver tube
    Max. Force 10,000xG
    Volume (Metric) 200 to 750μL
    For Use With Laboratory model microcentrifuges
    Membrane Material Cellulose Acetate
    Pore Size 0.2μm
  • Description & Specifications
Related Products
Product Content Feedback