Learn More
Thermo Scientific™ SUN-SRi™ High Recovery Vials
Combine large sample capacities while eliminating waste
Manufacturer:  Thermo Scientific™ 203 515
 View more versions of this product
Catalog No. 10-800-085
Add to cart
Description
Description
High Recovery vials feature a tapered base that acts to funnel the sample to a constricted center well where it can be extracted by most manual or automated methods. These vials are manufactured from Clear Type 1 Class A borosilicate glass or Amber glass with volumes from 1.5mL up to 60mL.
  • Available in amber or clear glass
  • Barcoding with linear labels or etching
  • Industry standard screw thread finish is compatible with open top or closed top caps
Order Info
Caps and septa sold separately.
Specifications
Specifications
Screw cap
High Recovery Vial
Clear
Borosilicate Glass
Tapered
45mm
A
15 mm
3.8 mL
1.02 dr.