Learn More
Thermo Scientific™ SUN-SRi™ High Recovery Vials
Combine large sample capacities while eliminating waste
$359.50
Specifications
Classification A
Closure Type Screw Cap
Material Borosilicate Glass
Shape Tapered
View More Specs
Products 3
Catalog Number Mfr. No. Color Volume (English) Price Quantity & Availability  
Catalog Number Mfr. No. Color Volume (English) Price Quantity & Availability  
10-800-082
View Documents Promotion Details
Thermo Scientific™
203 509
Amber 0.4 dr.
Pack of 100 for $359.50
Add to cart
10-800-090
View Documents Promotion Details
Thermo Scientific™
203 523
Clear 5.4 dr. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
High Recovery vials feature a tapered base that acts to funnel the sample to a constricted center well where it can be extracted by most manual or automated methods. These vials are manufactured from Clear Type 1 Class A borosilicate glass or Amber glass with volumes from 1.5mL up to 60mL.
  • Available in amber or clear glass
  • Barcoding with linear labels or etching
  • Industry standard screw thread finish is compatible with open top or closed top caps
Order Info
Caps and septa sold separately.
Specifications
Specifications
A
Screw Cap
Borosilicate Glass
Tapered