Learn More
Thermo Scientific™ SUN-SRi™ High Recovery Vials
Combine large sample capacities while eliminating waste
Manufacturer:  Thermo Scientific™ 203 511
 View more versions of this product
Catalog No. 10-800-083
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Screw cap
A
Clear
High Recovery Vial
44mm
0.4 dr.
1.5mL
13mm
Borosilicate Glass
1.5mL
Tapered