Learn More
Thermo Scientific™ SUN-SRi™ Luer-Slip Syringes
Siliconized and assembled with scale and tip shield.
Manufacturer:  Thermo Scientific™ 201 652
 View more versions of this product
Catalog No. 22-314387
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Syringe, Chemically resistant, inert polypropylene and polyethylene, Luer Slip, Bulk
  • Chemically resistant, inert polypropylene and polyethylene
  • No rubber plunger seals or silicone lubricants that may cause sample contamination
  • Bulk packaged, choice or sterile or nonsterile
  • LUER-Slip connection
Specifications
Specifications
Yes
Plastic
Sterile
Luer-slip Syringe
2 mL
Centered
Luer-slip
50 mL