Thermo Scientific™ SUN-SRi™ MicroMat CLR™ Silicone Sealing Mats
Superior resealability after up to 100 injections—resists coring and tearing
Manufacturer: Thermo Scientific™ 300002
 View more versions of this product
Catalog No. 14-823-248
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    No. per Case 10
    Well Shape Round
    Material Silicone
    No. of Wells 96
    Autoclavable Yes
    Product Type Sealing Mat
  • Description & Specifications
Related Products
Product Content Feedback