Learn More
Thermo Scientific™ SUN-SRi™ MicroMat™ PTFE-Coated Silicone Sealing Mats Available on GSA/VA Contract for Federal Government customers only.
Provides superior resealability after up to 100 injections, resists coring and tearing
Manufacturer:  Thermo Scientific™ 400 079
 View more versions of this product
Catalog No. 03-340-906
Add to cart
Description
Description
  • PTFE-coated silicone
  • Eliminates cross contamination of samples
  • Available pre-slit for easy penetration and to reduce vacuum
  • Dry heat autoclavable
  • Superior chemical compatibility
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Yes
TFE or Silicone
Flat Underside
10
Yes
Sealing Mat
Pink
96
Round