Learn More
Thermo Scientific™ SUN-SRi™ MicroMat™ PTFE-Coated Silicone Sealing Mats
Provides superior resealability after up to 100 injections, resists coring and tearing
$389.00 - $418.00
Specifications
Autoclavable Autoclavable
Type Sealing Mat
No. of Wells 96
No. per Case 10
View More Specs
Products 4
Catalog Number Mfr. No. Color Material Well Shape Description Price Quantity & Availability  
Catalog Number Mfr. No. Color Material Well Shape Description Price Quantity & Availability  
14-823-265
View Documents
Thermo Scientific™
400 073
Blue Silicone, PTFE Round Flat Underside
Case of 10 for $409.00
Add to cart
14-823-264
View Documents
Thermo Scientific™
400 067
Blue Silicone, PTFE Round Round Underside
Case of 10 for $418.00
Add to cart
14-823-258
View Documents
Thermo Scientific™
400 026
Blue Silicone, PTFE Square Flat Underside
Case of 10 for $401.50
Add to cart
03-340-906
View Documents
Thermo Scientific™
400 079
Pink TFE or Silicone Round Flat Underside
Case for $389.00
Add to cart
Description
Description
  • PTFE-coated silicone
  • Eliminates cross contamination of samples
  • Available pre-slit for easy penetration and to reduce vacuum
  • Dry heat autoclavable
  • Superior chemical compatibility
Specifications
Specifications
Autoclavable
Sealing Mat
96
10