Learn More
Thermo Scientific™ SUN-SRi™ MicroMat™ PTFE-Coated Silicone Sealing Mats
Provides superior resealability after up to 100 injections, resists coring and tearing
Supplier: Thermo Scientific™ 400 073
Description
- PTFE-coated silicone
- Eliminates cross contamination of samples
- Available pre-slit for easy penetration and to reduce vacuum
- Dry heat autoclavable
- Superior chemical compatibility
Specifications
Blue | |
Silicone, PTFE | |
96 | |
Sealing Mat |
Flat Underside | |
Autoclavable | |
10 | |
Round |